Learn More
Cole-Parmer™ Jenway™ Cuvette Holders
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
63/73/ 67 Series Spec
6850 Series Spectrophotometers
Pathlength:
10 to 100 mm
10 x 10 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Type | Cuvette Holder |
| For Use With (Equipment) | 6850 Series Spectrophotometers |
| Pathlength | 10 x 10 mm |