missing translation for 'onlineSavingsMsg'
Learn More

JMV 2959, MedChemExpress

MedChemExpress JMV 2959 is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32 nM.

Encompass_Preferred

153.90 € - 572.40 €

Specifications

Chemical Name or Material JMV 2959
CAS 925238-89-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H32N6O2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16484096
Encompass_Preferred
View Documents
Medchem Express
HY-U00433
25 mg
572.40 €
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16472638
Encompass_Preferred
View Documents
Medchem Express
HY-U00433
5 mg
153.90 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16460439
Encompass_Preferred
View Documents
Medchem Express
HY-U00433
10 mg
264.60 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

JMV 2959 is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32 nM.
Specifications

Specifications

JMV 2959
Off-White
C30H32N6O2
O=C(N[C@@H](C1=NN=C(CCC2=CC=CC=C2)N1CC3=CC=C(OC)C=C3)CC4=CNC5=C4C=CC=C5)CN
508.61
Research
Metabolism-protein/nucleotide metabolism
925238-89-7
Research
DMSO : 25 mg/mL (49.15 mM; ultrasonic and warming and heat to 60°C)
508.61
99.91%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
JMV 2959, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.