Learn More
behr Labor-Technik™ KEX 100 F Soxhlet Extractor
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
Brand:  behr Labor-Technik™ B00217706
Product Code. 10195450
  • 1175.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Compact equipment
    • specially developed lifting tube for homogeneous extraction cycles
      • the use of extractors with a stopcock does away with the need for any additional distillation after extraction (direct redirection of solvent to the reservoir)
Specifications
Specifications
Soxhlet Extractor
71.5 cm
7.5 kg
230 VAC
30 mL
23 cm
50/60 Hz
450 w