missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Aluminum Seal Caps
Click to view available options
Neck Finish:
11
Closure Liner:
PTFE/Natural Silicone Rubber
PTFE/Rubber
Color:
Natural
Red
No. per Case:
100 Cs.
1000 Cs.
Description
- Moderate to good resealability
- One piece
- PTFE-faced red rubber septa
- For general GC applications
- Moderate resealability
- Not recommended for multiple injections
- PTFE-faced natural rubber septa
- Good resealability
- Multiple injections
Specifications
Specifications
Color | Red |
For Use With (Application) | Multiple injection |
Material | Aluminum |
No. per Case | 1000 Cs. |
Neck Finish | 11 |
Closure Liner | PTFE/Rubber |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'