Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
35.11 € - 36.75 €
Specifications
For Use With Kimble-Chase Economy Columns
View More Specs
Products 3
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
13476349
View Documents
DWK Life Sciences
420404-1000
1 cm
36.75 €
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
11772404
View Documents
DWK Life Sciences
420404-2500
2.5 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11762404
View Documents
DWK Life Sciences
420404-1500
1.5 cm
36.50 €
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
Install on top of column packing to protect packing from disruption. Polyethylene disc is sealed to transparent acrylic ring.
Specifications
Specifications
Kimble-Chase Economy Columns