Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps
Product Code. 11892973
Click to view available options
Volume (Metric):
2 mL
4 mL
6 mL
8 mL
12 mL
16 mL
20 mL
24 mL
40 mL
Height (Metric):
28 mm
35 mm
38 mm
40 mm
45 mm
57 mm
60 mm
65 mm
70 mm
85 mm
95 mm
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
28 mm
Quantity:
144/Cs.
200/Cs.
288/Cs.
72/Cs.
Unit Size:
Pack of 144
Pack of 200
Pack of 288
Pack of 72
Product Code. 11892973
Brand: DWK Life Sciences 60811D1528
Please to purchase this item. Need a web account? Register with us today!
Packaged in corrugated trays with partitions
  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures
TRUSTED_SUSTAINABILITY
Specifications
Classification A
Material (Liner) Rubber
Color Clear
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 2 mL
Thread Style Screw
Volume (English) 0.5 dr.
Closure Material Phenolic
Closure Type Screw Cap (Closed-top)
Height (Metric) 28 mm
Quantity 200/Cs.