Learn More
DWK Life Sciences Kimble™ Kontes™ Pestle For All-Glass DUALL™ Tissue Grinders
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
Brand:  DWK Life Sciences 885451-0022
 View more versions of this product
Product Code. 10213202
  • 79.45 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
  • Combines both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
  • This ground glass model is ideal for connective tissue such as muscle, heart and lung
  • Construction is strong enough to allow the pestle to be motor-driven
  • Clearance between pestles and tubes is 0.004 in. to 0.006 in.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22
Tissue Grinder Pestle
Duall™ all Glass tissue grinders