Learn More
Macherey-Nagel™ KingFisher™ 24 Accessory Kit
Product Code. 15879167
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. 15879167
Brand: Macherey-Nagel™ 744953
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enables automated purification of genomic DNA using a NucleoMag™ kit on the KingFisher™ Flex platform for faster, high throughput performance with reliable yields.
  • Adapted NucleoMag™ procedure for instruments of the KingFisher™ Flex series
  • Reliable performance and excellent yields from different sample materials
  • Speeds up nucleic acid purification workflow: process 24 samples at once
  • Set includes Deep-well Plates and Tip Combs for processing in 24-well format
Specifications
For Use With (Equipment) KingFisher™ Flex platform, NucleoMag™ kit
For Use With (Application) Automated nucleic acid purification
Includes Deep-well Plates, Tip Combs for processing in 24-well format
Quantity 1 Set