Learn More
Klever Klever Excel Wide Cut
Product Code. p-7241158
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 50
Product Code. 17798987
Brand: Klever KCJ430B
Only null left
Add to basket
Add to basket
Recessed blade of the Klever Excel Wide Cut reduces cut injuries and damaged goods.
  • Cuts boxes, banding, film, tape, and thousands more applications.
  • Multiple position handle; two positions for control and reach.
  • Available in both narrow and wide heads with durable, high quality carbon steel blade.
  • Innovative plastic tape splitter.
  • Advanced Plastic Polymers.
  • Highest Safety Level.
  • Disposable.
Specifications
Product Type Klever Excel Wide Cutter
Disposable Single-use
Sterility Non-sterile
Color Blue