Learn More
DWK Life Sciences Kimble™ Kontes™ Cuvette Tube Washer for NMR Utility Washers
Replacement fask for utility washers.
Brand: DWK Life Sciences 459961-0000
This item is not returnable.
View return policy
Description
- Utility washers are used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items
- Unit is supplied with one gasket, a tube washer, an Erlenmeyer flask and a 3/8 in. hose connection
- Replacement flask, 1 EA
Specifications
Cuvette Washer | |
For NMR utility washer | |
Cuvette Washer |