missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Amino-N-Butyric acid, >98%, Thermo Scientific™

Product Code. p-7082774 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15825088

Brand: Thermo Scientific J11079.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1492-24-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00064415
InChI Key QWCKQJZIFLGMSD-VKHMYHEASA-N
Synonym l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
PubChem CID 80283
ChEBI CHEBI:35619
IUPAC Name (2S)-2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point >300°C
Packaging Plastic bottle
Linear Formula CH3CH2CH(NH2)COOH
Quantity 1 g
Beilstein 1720935
Formula Weight 103.12
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material L-alpha-Amino-N-Butyric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.