missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Carnosine, 98%

58.10 € - 1756.00 €

Chemical Identifiers

CAS 305-84-0
Molecular Formula C9H14N4O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00005207
InChI Key CQOVPNPJLQNMDC-ZETCQYMHSA-N
Synonym l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
PubChem CID 439224
ChEBI CHEBI:15727
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10311753
View Documents Promotion Details
Thermo Scientific Chemicals
208170010
1 g Glass bottle
58.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
10348440
View Documents Promotion Details
Thermo Scientific Chemicals
208170050
5 g Glass bottle
162.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10697684
View Documents Promotion Details
Thermo Scientific Chemicals
208170250
25 g Glass bottle
512.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
10239703
View Documents Promotion Details
Thermo Scientific Chemicals
208171000
100 g Glass bottle
1756.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

305-84-0
226.23
CQOVPNPJLQNMDC-ZETCQYMHSA-N
439224
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
C9H14N4O3
MFCD00005207
l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
CHEBI:15727
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN

Specifications

305-84-0
White
98%
MFCD00005207
15, 1851
+ 20.90
CQOVPNPJLQNMDC-ZETCQYMHSA-N
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
439224
226.23
Crystalline Powder
253.0°C
Authentic
C9H14N4O3
25, 516
+20.90° (20°C c=1.5,H2O)
l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
226.23
CHEBI:15727
98%
L-Carnosine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-169-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only