missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine disodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 15475339
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199329-53-8
Molecular Formula C6H12N2Na2O5S2
Molecular Weight (g/mol) 302.271
MDL Number MFCD00243291
InChI Key TVPDKJXPEMKMDM-BLZBDRIDSA-L
Synonym disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o
PubChem CID 71299670
IUPAC Name disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate
SMILES C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]

Specifications

Color White
Quantity 50 g
Formula Weight 302.27 (284.26 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material L-Cystine disodium salt monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'