Learn More
L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Product Code. 15478396
Click to view available options
Quantity:
250 mg
1 g
5 g
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4561-10-8
MDL Number MFCD00035103
Specifications
Color White
Molecular Formula C17H38ClN
Synonym tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride
InChI Key CEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-]
IUPAC Name trimethyl(tetradecyl)azanium;chloride
Molecular Weight (g/mol) 291.95
PubChem CID 20708
Formula Weight 363.84
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material L-Glutamic acid dibenzyl ester hydrochloride