missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid dimethyl ester hydrochloride, 99%, pure, Thermo Scientific Chemicals
47.60 € - 156.00 €
Chemical Identifiers
CAS | 23150-65-4 |
---|---|
Molecular Formula | C7H13ClNO4 |
Molecular Weight (g/mol) | 210.63 |
MDL Number | MFCD00038879 |
InChI Key | BPHCSYSXOKTCMA-UHFFFAOYNA-N |
Synonym | l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride |
PubChem CID | 12917568 |
IUPAC Name | dimethyl (2S)-2-aminopentanedioate;hydrochloride |
SMILES | [Cl].COC(=O)CCC(N)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11995381
|
Thermo Scientific Acros
441940100 |
10 g | N/A | ||||||
11925391
|
Thermo Scientific Acros
441940500 |
50 g |
156.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23150-65-4 | |
210.63 | |
BPHCSYSXOKTCMA-UHFFFAOYNA-N | |
12917568 | |
[Cl].COC(=O)CCC(N)C(=O)OC |
C7H13ClNO4 | |
MFCD00038879 | |
l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride | |
dimethyl (2S)-2-aminopentanedioate;hydrochloride |
Specifications
23150-65-4 | |
White | |
98.5% min. (Argentometry) | |
C7H13ClNO4 | |
26 | |
BPHCSYSXOKTCMA-UHFFFAOYNA-N | |
dimethyl (2S)-2-aminopentanedioate;hydrochloride | |
12917568 | |
99% | |
Powder |
91°C to 93°C | |
Authentic | |
Glass bottle | |
MFCD00038879 | |
l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride | |
[Cl].COC(=O)CCC(N)C(=O)OC | |
210.63 | |
211.65 | |
Pure | |
L-Glutamic acid dimethyl ester hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 245-461-
Product Identifier
- L-Glutamic acid dimethyl ester hydrochloride
RUO – Research Use Only