missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, 99%, Thermo Scientific Chemicals

Product Code. 11328805
Click to view available options
Quantity:
250 mg
1 g
5 g

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point ∽203°C (decomposition)
Beilstein 1721681
Merck Index 14,4739
Solubility Information Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol.
Formula Weight 119.12
Percent Purity 99%
Quantity 5 g
Optical Rotation −9° (c=2 in Water)
Chemical Name or Material L-Homoserine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'