missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Methionine, Cell Culture Reagent
25.80 € - 151.00 €
Chemical Identifiers
CAS | 63-68-3 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.21 |
MDL Number | MFCD00063097 MFCD00801344 |
InChI Key | FFEARJCKVFRZRR-BYPYZUCNSA-N |
Synonym | l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin |
PubChem CID | 6137 |
ChEBI | CHEBI:16643 |
SMILES | CSCC[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484389
|
Thermo Scientific Chemicals
J61904.18 |
50 g |
25.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15494389
|
Thermo Scientific Chemicals
J61904.22 |
100 g |
38.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15404399
|
Thermo Scientific Chemicals
J61904.36 |
500 g |
151.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63-68-3 | |
149.21 | |
FFEARJCKVFRZRR-BYPYZUCNSA-N | |
6137 | |
CSCC[C@H](N)C(O)=O |
C5H11NO2S | |
MFCD00063097 MFCD00801344 | |
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin | |
CHEBI:16643 |
Specifications
284°C (decomposition) | |
White | |
MFCD00063097 MFCD00801344 | |
14,5975 | |
Soluble in water | |
CSCC[C@H](N)C(O)=O | |
149.21 | |
CHEBI:16643 | |
Cell Culture Reagent | |
L-Methionine |
63-68-3 | |
C5H11NO2S | |
1722294 | |
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin | |
FFEARJCKVFRZRR-BYPYZUCNSA-N | |
(2S)-2-amino-4-methylsulfanylbutanoic acid | |
6137 | |
149.21 | |
Powder |
Safety and Handling
H500
missing translation for 'einecsNumber' : 200-562-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only