missing translation for 'onlineSavingsMsg'
Learn More

L-NIO dihydrochloride, Tocris Bioscience™

Product Code. 30001098
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NOS inhibitor

Inhibitor of nitric oxide (NO) synthase. Inhibits iNOS, eNOS and nNOS. Active in vivo.

Chemical Identifiers

CAS 159190-44-0
Molecular Formula C7H17Cl2N3O2
Molecular Weight (g/mol) 246.132
InChI Key RYCMAAFECCXGHI-ILKKLZGPSA-N
Synonym l-nio dihydrochloride, l-nio, 2hcl, l-n5-1-iminoethyl ornithine, dihydrochloride, n5-1-iminoethyl-l-ornithine dihydrochloride, l-nio hydrochloride, c7h15n3o2.2hcl, 2s-2-amino-5-1-aminoethylideneamino pentanoic acid dihydrochloride, 2s-2-amino-5-ethanimidamidopentanoic acid dihydrochloride
PubChem CID 2733507
IUPAC Name (2S)-2-amino-5-(1-aminoethylideneamino)pentanoic acid;dihydrochloride
SMILES CC(=NCCCC(C(=O)O)N)N.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 246.13
Percent Purity >98%
Chemical Name or Material L-NIO dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.