Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Product Code. 11765168
Click to view available options
For Use With:
CRS Rotor, L/S 14 Tubing
CRS Rotor, L/S 15 Tubing
CRS Rotor, L/S 16 Tubing
CRS Rotor, L/S 18 Tubing
CRS Rotor, L/S 24 Tubing
SS Rotor, L/S 14 Tubing
Unit Size:
Each
Product Code. 11765168
Brand: Masterflex™ 0701481
Please to purchase this item. Need a web account? Register with us today!
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
  • Replacement parts to keep pump head operating for an extended period of time
  • Two end bell assemblies are required for a complete head assembly
  • Choose replacement parts suitable for your I/P™ Standard pump head size
  • Suitable for use with L/S™ Standard Pump Heads
Specifications
For Use With CRS Rotor, L/S 14 Tubing
Material Polycarbonate
Size L/S™ Standard Pump Heads 07014-00, 07014-20, 07014-70, and 07014-72
Tubing Size L/S 14
Type Replacement End Bell Assembly