Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Product Code. p-7184897
Click to view available options
For Use With:
CRS Rotor, L/S 14 Tubing
CRS Rotor, L/S 15 Tubing
CRS Rotor, L/S 16 Tubing
CRS Rotor, L/S 18 Tubing
CRS Rotor, L/S 24 Tubing
SS Rotor, L/S 14 Tubing
Unit Size:
Each
Product Code. 14821302
Brand: Masterflex™ 0702481
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
  • Replacement parts to keep pump head operating for an extended period of time
  • Two end bell assemblies are required for a complete head assembly
  • Choose replacement parts suitable for your I/P™ Standard pump head size
  • Suitable for use with L/S™ Standard Pump Heads
Specifications
For Use With CRS Rotor, L/S 24 Tubing
Material Polycarbonate
Size L/S™ Standard Pump Heads 07024-00, 07024-20, 07024-70, and 07024-72
Tubing Size L/S 24
Type Replacement End Bell Assembly