missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Amyloid A Component Ab-1 Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1219-S0

 View more versions of this product

Product Code. 12669316

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: mcl

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Immunogen: Human amyloid A

Positive Control: Amyloid deposits in kidney

Cellular Localization: Extracellular

Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Amyloid A Component Ab-1
Monoclonal
Unconjugated
Human amyloid A
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
mcl
Mouse
100 μL
Neuroscience
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.