Learn More
Epredia™ Lab Vision™ Amyloid A Component Ab-1 Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1219-S1
Description
Host Species: Mouse
Clone: mcl
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Immunogen: Human amyloid A
Positive Control: Amyloid deposits in kidney
Cellular Localization: Extracellular
Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Amyloid A Component Ab-1 | |
Monoclonal | |
Unconjugated | |
Human amyloid A | |
RUO | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
mcl | |
Mouse | |
500 μL | |
Neuroscience | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.