missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-734-S0
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: 8A9
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta or seminoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Placental Alk. Phosphatase (PLAP) Ab-4 | |
Monoclonal | |
Unconjugated | |
Purified human Placental Alkaline Phosphatase (PLAP) | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
8A9 | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction