Learn More
Campingaz™ Labogaz 206 Bunsen Burner
Suitable for butane cartridge C 206.
Brand: Campingaz™ 17998
This item is not returnable.
View return policy
Description
- Burner with the Clip and Go™ assembly system for simple cartridge change
Specifications
Bunsen Burner | |
Power: 760w; Weight: 380g | |
1 Ea. |
Butane Cartridge C 206 | |
Clip and Go™ assembly system | |
380 g |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Campingaz™ Labogaz 206 Bunsen Burner
Spot an opportunity for improvement?Share a Content Correction