missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595850
This item is not returnable.
View return policy
Description
Positive Samples: Jurkat, HT-29, HeLa, U-251MG, mouse liver, mouse brain, mouse spleen, rat liver Immunogen sequence: AGVTASPPTD LIWKNQNSWG TGEDVKVILK NSQGEEVAQR STVFKTTIPE EEEEEEEAAG VVVEEELFHQ QGTPRASNRS CAIM.
Lamin B1 is a key member of the nuclear lamina structure, encoded by the LMNB1 gene. It is part of the B-type lamins, ubiquitously expressed across all mammalian cells, providing structural integrity and mechanical support by forming a meshwork along the inner nuclear membrane. Lamin B1 is involved in maintaining the shape and stability of the nucleus, playing a critical role in cellular processes such as genome organization, DNA replication, and transcription regulation. The lamin filaments are type V intermediate filament proteins, enabling the nucleus to withstand mechanical stress and preserve its functionality. Lamin B1's expression and dynamics vary during cell differentiation, influencing gene regulation. Its location within the euchromatin-enriched section of the genome suggests potential roles in chromatin organization and the epithelial-to-mesenchymal transition. Governed by complex interactions with other proteins, lamin B1 is integral to cellular architecture and health, with variations in its levels linked to developmental and disease processes. Recent studies have highlighted the importance of understanding the role and dynamics of lamin B1 in human-induced pluripotent stem cells and germ layer cells, offering insights into differentiation-dependent changes and their implications for nuclear structure.
Specifications
| Lamin B1 | |
| Polyclonal | |
| Unconjugated | |
| Lmnb1 | |
| 0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116685, 16906, 4001 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.03 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P14733, P20700, P70615 | |
| Lmnb1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 503-586 of human Lamin B1 (NP_0055641). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction