accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Laminated Polyester Fire Safety Signs: Directional Arrow - Right
Print
Brady™ Laminated Polyester Fire Safety Signs: Directional Arrow - Right
3.30 € - 4.90 €
Products
2
Description
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
10264794
View Documents
missing translation for 'viewProductCertificates'
Brady™
250514
100 x 100 mm
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10555694
View Documents
missing translation for 'viewProductCertificates'
Brady™
250516
200 x 200 mm
4.90 €
Each
Estimated Shipment: 10-12-2024
Log in
to see stock.
Add to basket
Description
Description
Videos
SDS
Documents
Documents
Product Certifications
Back to Top