Learn More
Brady™ Laminated Polyester Mandatory Face Protection Signs
Brand: Brady™ 222604
This item is not returnable.
View return policy
Specifications
Polyester | |
Permanent Acrylic | |
315 mm | |
-40°C to 120°C | |
Use Face Shield | |
White on Blue |
1 Ea. | |
Laminated Polyester | |
0.024 mm | |
Round | |
Face Protection Sign |