man
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Laminated Polyester Prohibitory Signs: Fire, Open Flame, and Smoking Prohibited
Print
Brady™ Laminated Polyester Prohibitory Signs: Fire, Open Flame, and Smoking Prohibited
3.20 € - 4.10 €
See promotional offers below
Specifications
Shape
Round
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
10090664
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Brady™
250003
100 mm
3.20 €
Each
View alternative products
Save Up to
»
In Stock
Log in
to see stock.
Add to basket
10325914
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Brady™
250004
200 mm
4.10 €
Each
View alternative products
Save Up to
»
In Stock
Log in
to see stock.
Add to basket
Specifications
Shape
Round
Videos
SDS
Documents
Documents
Product Certifications
Back to Top