missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ TR-FRET ERR alpha Coactivator Assay Kit, goat
LanthaScreen™ TR-FRET ERR alpha Coactivator Assay Kit, goat
Brand: Invitrogen™ PV4663
This item is not returnable.
View return policy
Description
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15130:The LanthaScreen™ ERR (estrogen related receptor) alpha TR-FRET Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential ERR alpha ligands as inverse agonists (sometimes referred to as antagonists) of coactivator displacement. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and an ERR alpha ligand-binding domain (ERR alpha-LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.
In the LanthaScreen™ ERR alpha TR-FRET Coactivator Assay, ERR alpha-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the ERR alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and displace coactivator peptide (Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET ERR alpha Coactivator Assay Kit contains ERR alpha-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Specifications
Tb (Terbium) | |
Microplate Reader | |
384-well Plate | |
Biochemical coregulator interaction | |
End Point | |
ESRRA, NR3B1, ERR1, ERR alpha | |
Co-Factor Interaction Assay, TR-FRET | |
TR-FRET ERR Alpha Coactivator Assay Kit |
Fluorescence | |
800 x 20 μL assays | |
Store in ultra-cold freezer (-68 to -85°C). | |
2101 | |
Dry Ice | |
LanthaScreen™ | |
ERR alpha |
For Research Use Only. Not for use in diagnostic procedures.