Learn More
Gilac™ Large Volume Container
Product Code. p-200009542
Click to view available options
Capacity (Metric):
100 L
310 L
500 L
Unit Size:
Each
Product Code. 17948521
Brand: Gilac™ G140521
Only null left
Add to basket
Add to basket
Large Volume Container has excellent mechanical and shock resistance.
  • Excellent mechanical resistance: made from high density polyethylene for increased shock resistance.
  • The durable, thick walls mean no distortion and warping.
  • Limited gaps and overlapping, equipped with edges so easy to handle.
Specifications
Type Storage Tote
Material HDPE
Color White
Shape Rectangular
Capacity (English) 26.4 gal.
Capacity (Metric) 100 L
Height (English) 2.2 ft.
Height (Metric) 665 mm