Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: L.pneumophila
Shop All Novacyt Group Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Description
- From a range of tests for the rapid detection of causative agents of respiratory illness.
- Specific tests available for a variety of infection detections.
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | L.. pneumophila |
| Type | Latex Agglutination Test |
| Quantity | 50 Tests |
| Test Time | 2 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering