missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Left Hinged Door for UN160plus

Product Code. 17569905 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN160plus Oven
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17569905

Brand: Memmert™ UN160plusB8

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Door hinged on the left for use with Memmert Ovens.

  • Hinged on the left
  • Accessory

Specifications

For Use With (Equipment) UN160plus Oven
Type Hinged Door
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.