missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LGALS1 (Human) Recombinant Protein

Human LGALS1 full-length ORF ( AAH01693, 1 a.a. - 135 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003956-P01.10ug

 View more versions of this product

Product Code. 16137371

  • 335.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Sequence: MACGLVASNLNLKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDFKIKCVAFD
Specifications

Specifications

AAH01693
lectin, galactoside-binding, soluble, 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LGALS1
GST
3956
Wheat germ expression system
10 μg
DKFZp686E23103, GAL1, GBP
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions