Documents
Product Information
Manuals
- 1
- 2
Support a diverse array of sample processing applications without the need to modify existing protocols.
Brand: Thermo Scientific™ 75006478
620.13 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Cryofuge 5500i series centrifuges
DoubleSpin Bucket | |
Double Spin Bucket | |
Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.