missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LH beta Antibody (SPM103), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11530JF646
This item is not returnable.
View return policy
Description
LH beta Monoclonal antibody specifically detects LH beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
LH beta | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
3972 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM103 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
Recombinant beta sub-unit of human LH beta (Uniprot: P01229) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LH beta Antibody (SPM103), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Spot an opportunity for improvement?Share a Content Correction