Learn More
Fisherbrand™ Lab Thermometer
Product Code. 10672272 Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
Subdivision:
0.5°C
1°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
155 mm
305 mm
Unit Size:
Each
Product Code. 10672272
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A general purpose/stirring thermometer for a wide application of laboratory needs.
  • With amber graduations
  • Non-roll fitting
  • Reinforced bulb for stirring
Specifications
For Use With (Application) General use
Immersion Type Total Immersion
Length (Metric) 305 mm
Thermometer Fill Red Spirit
Product Type Liquid Filled Thermometer
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -20°C
Temperature Range (Metric) -20°C to +110°C
Subdivision 1°C