Learn More
Borer Chemie™ deconex™ 11 UNIVERSAL Liquid
For the manual and automatic cleaning of laboratory glassware, instruments and utensils. Borer Chemie™ deconex™ 11 UNIVERSAL Liquid provides residue-free removal of stubborn organic and inorganic soil.
Brand:  Borer Chemie™ 500100.00-F10W
Product Code. 10712721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Environmentally friendly
  • Mildly alkaline with phosphates
  • With active chlorine
  • Also for radioactive decontamination
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Borer Chemie™ deconex™ 11 UNIVERSAL Liquid > Quantity: 1kg