Learn More
Lithium acetylide ethylenediamine complex, tech. 85%, Thermo Scientific Chemicals
65.10 € - 251.25 €
Chemical Identifiers
CAS | 6867-30-7 |
---|---|
Molecular Formula | C4H9LiN2 |
Molecular Weight (g/mol) | 92.07 |
MDL Number | MFCD00013183 |
InChI Key | QJQWXTYPTBEPGS-UHFFFAOYSA-N |
Synonym | lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine |
PubChem CID | 2724010 |
IUPAC Name | lithium;ethane-1,2-diamine;ethyne |
SMILES | [Li]C#C.NCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456780
|
Thermo Scientific Alfa Aesar
L12401.14 |
25 g | N/A | ||||||
11368785
|
Thermo Scientific Alfa Aesar
L12401.22 |
100 g | N/A | ||||||
11466780
|
Thermo Scientific Alfa Aesar
L12401.22 |
100 g |
251.25 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.
Solubility
Soluble in ethylene diamine, n-propyl amine, n-butyl amine, pyridine, dimethyl formamaide, diglyme, dioxan and dimethylsulfoxide.
Notes
Moisture and light sensitive. Store in cool place. Incompatible with alcohols, acids, oxidizing agents, water. It reacts with water to form lithium hydroxide and acetylene, which is a flammable gas. Over time, pressure may increase causing containers to burst. Handle and open container with care.
Chemical Identifiers
6867-30-7 | |
92.07 | |
QJQWXTYPTBEPGS-UHFFFAOYSA-N | |
2724010 | |
[Li]C#C.NCCN |
C4H9LiN2 | |
MFCD00013183 | |
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine | |
lithium;ethane-1,2-diamine;ethyne |
Specifications
6867-30-7 | |
1.04 | |
C4H9LiN2 | |
MFCD00013183 | |
3593824 | |
lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine | |
QJQWXTYPTBEPGS-UHFFFAOYSA-N | |
lithium;ethane-1,2-diamine;ethyne | |
2724010 | |
90% | |
Lithium acetylide ethylenediamine complex |
∽76°C (decomposition) | |
Amine-like | |
HC≡CLi·NH2CH2CH2NH2 | |
UN3131 | |
Moisture sensitive | |
Soluble in ethylene diamine,n-propyl amine,n-butyl amine,pyridine,dimethyl formamaide,diglyme,dioxan and dimethylsulfoxide. | |
[Li]C#C.NCCN | |
92.07 | |
92.07 | |
Technical |
Safety and Handling
GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H260-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 229-967-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.