missing translation for 'onlineSavingsMsg'
Learn More
Lithium periodate dihydrate, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11328118

Lithium periodate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11328118
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11328118

Brand: Thermo Scientific Alfa Aesar 039355.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307310-74-3
Molecular Formula ILiO4
Molecular Weight (g/mol) 197.84
MDL Number MFCD00049474
InChI Key SYWXNZXEJFSLEU-UHFFFAOYSA-M
Synonym lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6
PubChem CID 23711816
IUPAC Name lithium;periodate
SMILES [Li+].[O-]I(=O)(=O)=O

Specifications

Melting Point >370°C
Physical Form Powder
Quantity 50 g
UN Number UN1479
Solubility Information Soluble in water.
Formula Weight 233.87 (197.84 Anhydrous)
Grade Reagent
Odor Odorless
Chemical Name or Material Lithium periodate dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT11K015 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number039355.30,039355.18
Lot NumberT27J001 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number039355.30,039355.18
Lot NumberT22I028 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number039355.30,039355.18
Lot NumberJ12U034 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039355.30,039355.18
Lot NumberL27Q043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039355.30,039355.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium periodate dihydrate, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.