missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Load 1 Protein-Gel Loading Buffer
POA
Specifications
Concentration | 4X |
---|---|
Product Type | Protein Sample Loading Buffer |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10317803
|
Carl Roth™
K929.2 |
4 x 10 mL |
POA
40mL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10155103
|
Carl Roth™
K929.1 |
10 mL |
POA
10mL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4X | |
Protein Sample Loading Buffer | |
Liquid |