Learn More
Lenz Laborglasinstrumente™ Delivery Adapter, Long, Bent
For feeding distillate from condensers into receiving flasks. Lenz Laborglasinstrumente™ Delivery Adapters, Long, Bent are manufactured from durable DURAN™ glass tubing.
21.00 € - 23.00 €
Specifications
Type Long Bent Delivery Adapter
Material Duran Glass
View More Specs
Products 4
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
11816164
View Documents
Lenz Laborglasinstrumente™
05263001
NS 14/23
21.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11896074
View Documents
Lenz Laborglasinstrumente™
05263002
NS 19/26
21.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11876124
View Documents
Lenz Laborglasinstrumente™
05263003
NS 24/29
23.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11826114
View Documents
Lenz Laborglasinstrumente™
05263004
NS 29/32
23.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • With NS socket
  • Long bent tube with angle of 105°
  • 12mm diameter
Specifications
Specifications
Long Bent Delivery Adapter
Duran Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Delivery Adapter, Long, Bent