Learn More
Hamilton™ Long Life Instrument Syringes
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
150 μL
250 μL
500 μL
1 mL
5 mL
Unit Size:
Each
Description
- Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
- Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
Specifications
| Material | Borosilicate Glass, PTFE, UHMWPE |
| Diameter (Metric) Outer | 9.02 mm |
| Quantity | 1 Ea. |
| Volume (Metric) | 500 μL |
| Connection Type | Luer Lock |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Comparta una corrección de contenido