missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System Recovery Cassettes
Eliminates agarose gel preparation, band excision and purification. Lonza™ FlashGel™ - System Recovery Cassettes deliver recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.
330.91 €
Specifications
Product Type | System Recovery Cassettes |
---|---|
Wells | 8 + 1-well |
For Use With (Application) | Subsequent re-amplification, cloning or other techniques |
Kit Contents | Agarose Gel, Stain and Buffer |
Material | Agarose |
Product Code | Brand | Formulation | Separation Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Separation Range | Price | Quantity & Availability | ||||
11557865
|
Lonza™
57051 |
1.2% Agarose | 50 to 10,000 bp |
330.91 €
Pack of 9 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11592106
|
Lonza™
57022 |
2.2% Agarose | 10 to 1000 bp |
330.91 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
- Recover samples directly, without band excision or purification
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
System Recovery Cassettes | |
Subsequent re-amplification, cloning or other techniques | |
Agarose | |
18° to 26°C | |
2.7 in. | |
7 cm | |
10.7 cm | |
0.6 in. |
8 + 1-well | |
Agarose Gel, Stain and Buffer | |
9 Pack | |
4.5 in. | |
11.5 cm | |
4.2 in. | |
8.4 cm | |
1.7 cm |
Safety and Handling
missing translation for 'shelfLife' : 5 Months