Learn More
Lonza™ FlashGel™ System Sample Preparation Reagents
Product Code. 11537805
Click to view available options
Quantity:
2 x 500 μL
5 x 1 mL
Unit Size:
Pack of 2
Pack of 5
Product Code. 11537805
Brand: Lonza™ 50462
Please to purchase this item. Need a web account? Register with us today!
For use with the Lonza FlashGel System. FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.
  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
Product Type DNA Loading Dye
Form Liquid
Quantity 5 x 1 mL
Storage Requirements Room Temperature