Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
Brand:  Lonza™ 54711
 View more versions of this product
Product Code. 11507765
  • 783.64 € / 50 sheets
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
  • Covalently attaches to the acrylamide monomers during the polymerization reaction
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining, destaining, and drying
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13.8 x 15.8 cm
Polyester
0.007 in.
6.2 in.
5.4 in.
PAG Film for Acrylamide Gels
50/Pk.
0.02 cm
15.8 cm
15.8 cm