Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
Brand:  Lonza™ 54727
 View more versions of this product
Product Code. 11527765
  • 1075.00 € / 50 sheets
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Covalently attaches to the acrylamide monomers during the polymerization reaction
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining, destaining, and drying
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12.4 x 25.8 cm
Polyester
0.007 in.
10.1 in.
4.8 in.
PAG Film for Acrylamide Gels
50/Pk.
0.02 cm
25.8 cm
25.8 cm