Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
Brand:  Lonza™ 54729
 View more versions of this product
Product Code. 11537765
  • 1240.00 € / 50 sheets
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Covalently attaches to the acrylamide monomers during the polymerization reaction
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining, destaining, and drying
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16.5 x 22 cm
Polyester
0.007 in.
10.1 in.
6.4 in.
PAG Film for Acrylamide Gels
50/Pk.
0.02 cm
22 cm
22 cm