Learn More
SeaKem™ HE Agarose, Lonza™
  • For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
  • High EEO (0.23 –0.26)
  • Standard gelling temperature agarose
Brand:  Lonza 50020
 View more versions of this product
Product Code. 11527775
  • 769.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
SeaKem HE Agarose
125 g