man
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Reagents and Buffers
Electrophoresis Gel Casting Reagents
SeaKem™ HEEO Agarose, Lonza™
Print
SeaKem™ HEEO Agarose, Lonza™
208.00 € - 969.00 €
See promotional offers below
Specifications
Chemical Name or Material
SeaKem HEEO Agarose
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Quantity
Price
Quantity & Availability
Product Code
Brand
Quantity
Price
Quantity & Availability
11567775
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50031
25 g
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11557775
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50030
125 g
969.00 €
Each
Estimated Shipment: 10-01-2025
Log in
to see stock.
Add to basket
Specifications
Chemical Name or Material
SeaKem HEEO Agarose
Videos
SDS
Documents
Documents
Product Certifications
Back to Top