Learn More
SeaPlaque™ Agarose, Lonza™
Product Code. p-2861590
Click to view available options
Quantity:
25 g
125 g
Unit Size:
125g
Each
Product Code. 11587785
Brand: Lonza 50101
Please to purchase this item. Need a web account? Register with us today!
  • For preparative DNA and RNA electrophoresis
  • Molecular biology grade
  • ≤0.10 EEO
  • Low melting temperature argarose (26°–30°C Gelling temperature)
  • DNase/RNase-free
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SeaPlaque™ Agarose
Quantity 25 g